Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CTXB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12862R-HRP
BOSSBS-12862R-HRPEA 883 EUR InStock
BOSSBS-12862R-HRP
Anti-CTXB Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The B subunit of cholera toxin (CtxB) binds to a GM1-ganglioside receptor, a ubiquitous glycolipid cell surface receptor. This binding is widely accepted to initiate toxin action by triggering uptake and delivery of the toxin A subunit into cells. The beta chain has no toxic activity by itself. The holotoxin consists of a pentameric ring of B subunits whose central pore is occupied by the A subunit. The A subunit contains two chains, A1 and A2, linked by a disulfide bridge. The A subunit (and Cholera toxin) activates the adenylate cyclase enzyme in cells of the intestinal mucosa leading to increased levels of intracellular cAMP.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CTXB
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge