Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ABTB1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12881R-CY5
BOSSBS-12881R-CY5EA 900 EUR InStock
BOSSBS-12881R-CY5
Anti-ABTB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
ABTB1 is a 478 amino acid protein localized to the cytoplasm. ABTB1 contains two ANK repeats and two BTB (POZ) domains. The BTB (POZ) domain is thought to be involved in protein-protein interactions, and may indicate a role of ABTB1 in developmental processes. It has also been suggested that ABTB1 may be a mediator of the PTEN growth-suppressive signaling pathway. ABTB1 is ubiquitously expressed in all fetal tissues, with lower levels of expression found in adult heart. ABTB1 exists as four isoforms produced by alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABTB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge