Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BPY2B Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12882R-A555
BOSSBS-12882R-A555EA 893 EUR InStock
BOSSBS-12882R-A555
Anti-BPY2B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Mammalian spermatogenesis is a complex developmental process. Mutations at multiple loci and in structurally and functionally disparate genes in the genome affect gametogenesis. The analysis of mutations has provided insight into biochemical pathways required for completion of this process. The basic protein on Y chromosome 2 gene (BPY2, also designated VCY2) is located in a frequently deleted azoospermia factor c region. Three copies (paralogs) of the BPY2 gene (BPY2A, BPY2B, BPY2C) reside in the AZFc region. BPY2 protein expression is localized to the nuclei of spermatogonia, spermatocytes and round spermatids, but is absent from elongated spermatids. Impaired expression of BPY2 in infertile men suggests its involvement in male germ cell development. BPY2 interacts with MAP-1S, which shares homology with microtubule-associated proteins (MAPs), suggesting a role for BPY2 within the cytoskeletal network.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPY2B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge