Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BRAF35 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12883R-A680
BOSSBS-12883R-A680EA 890 EUR InStock
BOSSBS-12883R-A680
Anti-BRAF35 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The breast cancer susceptibility gene (BRCA1) localizes to chromosome 17q. Mutations within this gene account for approximately 45% of families with high incidence of breast cancer and at least 80% of families with increased incidence of both early-onset breast cancer and ovarian cancer. A second breast cancer susceptibility gene, BRCA2, located on chromosome 13q12-13, also confers a high incidence of breast cancer, but unlike BRCA1, BRCA2 does not confer a substantially elevated risk of ovarian cancer. The BRCA2-Associated Factor 35 (BRAF35) protein forms a complex with BRCA2, which associates with condensed chromatin during histone H3 phosphorylation. BRAF35 expression levels are highest in proliferating tissues and parallel BRCA2 expression patterns. The structure of BRAF35 includes a kinesin-like coiled coil domain and a nonspecific DNA binding HMG domain. The chromatin localisation of BRAF35 and antibody microinjection studies indicate a role for the BRAF35/BRCA2 complex in cell cycle regulation.

Type: Primary
Antigen: BRAF35
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-100/317
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge