Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HMG20B Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12883R-CY3
BOSSBS-12883R-CY3EA 900 EUR InStock
BOSSBS-12883R-CY3
Anti-HMG20B Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The breast cancer susceptibility gene (BRCA1) localizes to chromosome 17q. Mutations within this gene account for approximately 45% of families with high incidence of breast cancer and at least 80% of families with increased incidence of both early-onset breast cancer and ovarian cancer. A second breast cancer susceptibility gene, BRCA2, located on chromosome 13q12-13, also confers a high incidence of breast cancer, but unlike BRCA1, BRCA2 does not confer a substantially elevated risk of ovarian cancer. The BRCA2-Associated Factor 35 (BRAF35) protein forms a complex with BRCA2, which associates with condensed chromatin during histone H3 phosphorylation. BRAF35 expression levels are highest in proliferating tissues and parallel BRCA2 expression patterns. The structure of BRAF35 includes a kinesin-like coiled coil domain and a nonspecific DNA binding HMG domain. The chromatin localization of BRAF35 and antibody microinjection studies indicate a role for the BRAF35/BRCA2 complex in cell cycle regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMG20B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge