Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BRD2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12888R
BOSSBS-12888REA 725 EUR InStock
BOSSBS-12888R
Anti-BRD2 Rabbit Polyclonal Antibody
Antikörper
The bromodomain-containing proteins include BRD2, BRD3, BRD4 and BRDT. BRD2 (RING3 protein) is a mitogen-activated nuclear protein whose gene is located in the human MHC II region, suggesting its relation to HLA-associated diseases. The gene encoding BRD3 (RING3-like protein) contains two bromodomains and maps to chromosome 9q34. BRD4 (HUNK1 protein) is a nuclear protein involved in the regulation of chromosomal dynamics during mitosis. The testis-specific bromodomain protein BRDT contains a PEST sequence, indicating that it undergoes rapid intracellular degradation. The bromodomain-containing proteins are ubiquitously expressed.

  • WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: BRD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge