Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CYP2A6 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12923R-CY7
BOSSBS-12923R-CY7EA 900 EUR InStock
BOSSBS-12923R-CY7
Anti-CYP2A6 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
P450 enzymes constitute a family of monooxygenase enzymes that are involved in the metabolism of a wide array of endogenous and xenobiotic compounds (1). Several P450 enzymes have been classified by sequence similarities as members of the CYP1A and CYP2A subfamilies (2). NADPH cytochrome P450 reductase is a microsomal enzyme responsible for the transfer of electrons from NADPH to cytochrome P450 enzymes during the P450 catalytic cycle (3,4). NADPH cytochrome P450 reductase is localized to the endoplasmic reticulum where it is also able to transfer electrons to heme oxygenase and cytochrome b5 (5,6). NADPH cytochrome P450 reductase is structurally related to two separate flavoprotein families, ferredoxin nucleotide reductase (FNR) and flavodoxin (7). Electron transfer of NADPH cytochrome P450 reductase requires the binding of two flavin cofactors, FAD and FMN, to the FNR and flavodoxin domains, respectively (8).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP2A6
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge