Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CYP20A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12927R-A488
BOSSBS-12927R-A488EA 893 EUR InStock
BOSSBS-12927R-A488
Anti-CYP20A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Cytochrome P450 proteins are heme-thiolate monooxygenases that mediate NADPH-dependent electron transport and function to oxidize a variety of structurally unrelated compounds, including steroids, fatty acids and xenobiotics. Specifically, Cytochrome P450s are responsible for metabolizing arachidonic acid to hydroxyeicosatetraenoic acid (a regulator of blood pressure) and epoxyeicosatrienoic acid (a molecule involved in signaling events). CYP20A1 (cytochrome P450, family 20, subfamily A, polypeptide 1), also known as CYP-M, is a 462 amino acid single-pass membrane protein that belongs to the cytochrome P450 family. CYP20A1 is thought to carry its own oxygen as it lacks a conserved I-helix motif and one amino acid of its conserved heme binding site.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP20A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge