Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CTRB1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12933R-CY3
BOSSBS-12933R-CY3EA 900 EUR InStock
BOSSBS-12933R-CY3
Anti-CTRB1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Chymotrypsins are digestive enzymes that can perform proteolysis by cleaving peptides at the carboxyl side of tyrosine, tryptophan, leucine and phenylalanine, although over time they can also hydrolyze other amide bonds, especially those with leucine-donated carboxyls. Chymotrypsins cleave peptide bonds by attacking the non-reactive carbonyl group with a powerful nucleophile, which momentarily becomes covalently bonded to the substrate to form an intermediate. Chymotrypsin B (CTRB1) and Chymotrypsin B2 (CTRB2) are synthesized in the pancreas by protein biosynthesis as precursors that are enzymatically inactive, but become active as three polypeptide molecules that are interconnected by disulfide bonds.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTRB1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge