Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CTBS Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12937R-A350
BOSSBS-12937R-A350EA 893 EUR InStock
BOSSBS-12937R-A350
Anti-CTBS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
CTBS is an evolutionarily conserved member of the glycosyl hydrolase 18 family of proteins. Localizing to the lysosome, CTBS plays a role in the degradation of asparagine-linked (Asn-linked) glycoproteins. Glycoproteins are translocated to lysosomes via endocytosis or autophagy where they are broken down by proteases and glycosidases. The catabolism of glycoproteins is an important step in the regular turnover of cellular contents and in maintaining the homeostasis of glycosylation. CTBS functions as a glycosidase that cleaves the reducing end GlcNAc from the core chitobiase unit of oligosaccharides. Before this reaction can occur, AGA (the lysosomal glycosylasparaginase) must first remove the Asn from the Asn-linked glycoprotein to expose the reducing end GlcNAc, thereby allowing CTBS to access the exposed moiety.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTBS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge