Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CSK Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12941R-CY3
BOSSBS-12941R-CY3EA 900 EUR InStock
BOSSBS-12941R-CY3
Anti-CSK Rabbit Polyclonal Antibody (Cy3®)
Antikörper
All members of the Src gene family of tyrosine kinases are characterized by a carboxy terminal domain tyrosine which is highly phosphorylated in the inactive form of the enzyme and phosphorylated to a much lesser extent when the enzyme is active. In the case of Src p60, Y527 is this tyrosine; however, a mutant form of c-Src in which Y527 is replaced by phenylalanine is transforming and displays 5- to 10-fold elevated kinase activity compared to its normal counterpart. Csk has been identified as a Src-related tyrosine kinase having both SH2 and SH3 domains and a catalytic domain but lacking sequences amino terminal to the SH3 domain as well as carboxy terminal regulatory sequences. Csk phosphorylates Src on Y527 and also downregulates Lyn, Fyn and Lck by tyrosine phosphorylation of carboxy terminal regulatory sites.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSK
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge