Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CSRP2BP Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12947R
BOSSBS-12947REA 725 EUR InStock
BOSSBS-12947R
Anti-CSRP2BP Rabbit Polyclonal Antibody
Antikörper
CRP2BP is a 782 amino acid protein encoded by the human gene CSRP2BP. CRP2BP specifically interacts with the double LIM domain protein CRP2. The LIM domain is a conserved cysteine and histidine-containing structural module of two tandemly arranged zinc fingers. It has been identified in single or multiple copies in a variety of regulatory proteins, either in combination with defined functional domains, like homeodomains, or alone, like in the CRP family of LIM proteins. Members of the cysteine- and glycine-rich protein family (CRP1, CRP2 and CRP3) contain two zinc-binding LIM domains, LIM1 (amino-terminal) and LIM2 (carboxyl-terminal), and are implicated in diverse cellular processes linked to differentiation, growth control and pathogenesis. Although present in cytoplasm, CRP2BP is mainly a ubiquitously expressed nuclear protein, with highest expression in skeletal muscle and heart.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CSRP2BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge