Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRTAC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12948R-A488
BOSSBS-12948R-A488EA 893 EUR InStock
BOSSBS-12948R-A488
Anti-CRTAC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
CRTAC1 is a 661 amino acid secreted protein. CRTAC1 is O-glycosylated and contains one EGF-like domain and four FG-GAP repeats. Up-regulated in mesenchymal stem cells undergoing chondrogenic differentiation and also by growth factor BMP-4, CRTAC1 is found in the interterritorial matrix of articular deep zone cartilage. CRTAC1 is expressed as three isoforms produced by alternative splicing events. All isoforms are expressed in bone, cartilage and lung, while isoforms 1 and 2 are expressed in the brain. The gene that encodes CRTAC1 maps to human chromosome 10, which contains over 800 genes and 135 million nucleotides, making up nearly 4.5% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRTAC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge