Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRTAP Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12949R-A647
BOSSBS-12949R-A647EA 893 EUR InStock
BOSSBS-12949R-A647
Anti-CRTAP Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
CRTAP is a secreted protein localizing to the extracellular space that plays a role in collagen post-translational modifications, extracellular fibril assembly and intracellular trafficking. CRTAP is widely expressed with predominant expression in articular chondrocytes. It contains a signal peptide and a tetratricopeptide-like helical domain and is essential for normal bone formation. In the endoplasmic reticulum (ER), CRTAP forms a complex with Gros1 and CyPB (cyclophilin B) and is required for the efficient 3-hydroxylation of target prolyl residues in Collagen Type I molecules, the major structural proteins of skin and bone. Mutations in the gene encoding CRTAP can lead to autosomal recessive osteogenesis imperfecta (OI) type 7 and type 2B. OI, also known as brittle bone disease, is characterized by bone fragility and susceptibility to fractures. OI type 7 is a mild form of this disorder, while OI type 2B is a neonatal lethal condition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRTAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge