Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CRTC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12950R-FITC
BOSSBS-12950R-FITCEA 900 EUR InStock
BOSSBS-12950R-FITC
Anti-CRTC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
TORC3 is a 619 amino acid protein that localizes to both the cytoplasm and the nucleus and belongs to the TORC family. Expressed in lung tissue and B and T lymphocytes, as well as in colon, brain, ovary, kidney, prostate, colon and heart, TORC3 functions as a transcriptional coactivator for CREB-1, thereby regulating the expression of CREB-activated genes, and is also thought to activate the SIK/TORC signaling pathway. TORC3 exists as multiple alternatively spliced isoforms and, in addition to its role in transcriptional activation, is thought to induce mitochondrial biogenesis, specifically in muscle cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRTC3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge