Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ASAH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12976R-HRP
BOSSBS-12976R-HRPEA 900 EUR InStock
BOSSBS-12976R-HRP
Anti-ASAH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Acid ceramidase catalyzes the degradation of ceramide in normal tissues, and deficiency leads to accumulation of ceramide in tissues, a hallmark of Farber disease. Effected individuals experience early onset joint problems and neurological problems, owing to mutations in the acid ceramidase gene. Bioinformatic analysis of gene expression also reveals acid ceramidase to be among the 5 most important genes associated with melanoma. In addition to ceramide hydrolysis, purified acid ceramidase also exhibits the ability to catalyze ceramide synthesis, utilizing [14C]lauric acid and sphingosine as substrates. Interestingly, pH regulates which reaction is favored; for hydrolysis the pH optimum is 4.5, whereas for the reverse reaction favors a pH of 5.5, further supporting a complex and central role for acid ceramidase in sphingolipid metabolism.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ASAH1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge