Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DAPP1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12983R
BOSSBS-12983REA 743 EUR InStock
BOSSBS-12983R
Anti-DAPP1 Rabbit Polyclonal Antibody
Antikörper
B cell adapter molecule is also designated dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide (DAPP1) or B lymphocyte adapter protein. BAM32 is a B cell-associated adapter that is crucial for B cell antigen receptor signaling regulation. BAM32 interacts with Ptdlns and PLC g2 and, upon B cell activation, the protein is phosphorylated on tyrosine residues. It is a mainly cytoplasmic protein that can translocate to the cell membrane after cell stimulation. BAM32, which contains one PH domain and one SH2 domain, is primarily expressed in placenta and lung tissues, but can also be detected in heart, liver, pancreas and brain.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DAPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge