Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DKC1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13007R-CY3
BOSSBS-13007R-CY3EA 900 EUR InStock
BOSSBS-13007R-CY3
Anti-DKC1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Dyskerin (NAP57) associates with the chaperone protein Nopp140 and forms a small ribonucleoprotein particle with GAR1 (NOLA1), NHP2 (NOLA2) and Nop10 for the isomerization of uridine to pseudouridine (1). GAR1, NHP2 and dyskerin localize to the dense fibrillar component of the nucleolus and in nuclear Cajal bodies (2). The dyskerin gene maps to chromosome Xq28 (3). Missense mutations in the dyskerin gene interfere with normal nuclear localization of dyskerin and cause Dyskeratosis congenita (DKC) (4). DKC is a rare, X-linked bone marrow disorder characterized by cutaneous hyperpigmentation, dystrophy of the nails, atrophy of the testicles and leukoplakia of the oral mucosa. The GAR1 gene maps to chromosome 4q25 (5,6). The NHP2 gene maps to chromosome 5q35.3 and encodes a 155-amino acid protein (2,7).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DKC1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge