Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-POLI Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13018R-CY3
BOSSBS-13018R-CY3EA 900 EUR InStock
BOSSBS-13018R-CY3
Anti-POLI Rabbit Polyclonal Antibody (Cy3®)
Antikörper
DNA polymerase activity is essential for replication, repair, recombination and mutagenesis. DNA polymerases can often bypass DNA lesions that block DNA replication, thereby allowing the replication of damaged DNA. One such DNA polymerase is the distributive enzyme DNA Pol i, which is encoded by the POLI gene. POLI is located on human chromosome 18q21.2, a region often implicated in the etiology of many human cancers. At thymine templates, DNA Pol i is highly error-prone when replicating undamaged DNA in that it favors the misincorporation of guanine over the correct nucleotide, adenosine. DNA Pol i also promotes the replication of damaged DNA by misincorporating deoxynucleotides opposite DNA lesions. DNA Pol i acts sequentially with DNA Pol Ω, which is essential for damage-induced mutagenesis, to complete the DNA lesion bypass. Therefore, replication involving DNA Pol i is likely to be highly mutagenic.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POLI
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge