Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DNA polymerase mu Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13021R-A750
BOSSBS-13021R-A750EA 890 EUR InStock
BOSSBS-13021R-A750
Anti-DNA polymerase mu Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
DNA polymerase mu shares a number of characteristics with DNA polymerase Beta as well as with terminal deoxynucleotideyltransferase. Pol mu purportedly plays a role in microhomology mediated joining and the repair of double-stranded breaks. However, unlike other DNA polymerases, which show substrate specificity for deoxynucleotides, DNA Pol mu incorporates both deoxynucleotides and ribonucleotides in a template- directed manner. This unusual capability implies a novel role for this polymerase in DNA repair.

Type: Primary
Antigen: DNA polymerase mu
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 261-360/494
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge