Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DNMT3L Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13026R-HRP
BOSSBS-13026R-HRPEA 900 EUR InStock
BOSSBS-13026R-HRP
Anti-DNMT3L Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Methylation at the 5'-position of cytosine is the only known naturally occurring covalent modification of the mammalian genome. DNA methylation requires the enzymatic activity of DNA 5-cytosine methyltransferase (Dnmt) proteins, which catalyze the transfer of a methyl group from S-adenosyl methionine to the 5'-position of cytosines, thereby repressing expression of the target gene. Dnmt3L (DNA (cytosine-5)-methyltransferase 3-like) is a 387 amino acid protein that contains one ADD-type zinc finger and is a member of the Dnmt family. Localized to the nucleus and expressed at lows levels in thymus, testis and ovary, Dnmt3L does not exhibit DNA methyltransferase activity, but is able to stimulate de novo methylation by Dnmt3 and is thought to play a key role in the establishment of genomic imprints. Additionally, Dnmt3L interacts with histone deacetylase 1 (HDAC1) and, through this interaction, mediates transcriptional repression. Multiple isoforms of Dnmt3L exist due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DNMT3L
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge