Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZBED1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13030R-A488
BOSSBS-13030R-A488EA 1350 EUR InStock
BOSSBS-13030R-A488
Anti-ZBED1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ZBED1 is a 694 amino acid protein that localizes specifically to granular structures within the nucleus. Expressed ubiquitously at low levels and present at higher levels in heart, placenta, spleen and skeletal muscle, ZBED1 is thought to function as a transcription factor that regulates a number of ribosomal protein (RP) encoding genes, thereby playing a role in the cell cycle and in cell proliferation events. ZBED1 contains one BED-type zinc finger and binds specifically to 5'-TGTCG[CT]GA[CT]A-3' DNA regions found in RP promotors. Additionally, ZBED1 binds strongly to the promotor region of Histone H1 (a protein required for the condensation of nucleosomes into higher order structures), subsequently activating H1 transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBED1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge