Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DRIL1/ARID3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13031R-A680
BOSSBS-13031R-A680EA 890 EUR InStock
BOSSBS-13031R-A680
Anti-DRIL1/ARID3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ARID3A, also known as DRIL1 in humans and Bright (for B cell regulator of IgH transcription) in mice, are the mammalian homologs of the Drosophila Dri (dead ringer) protein. ARID3A is developmentally regulated and is expressed in a restricted set of cells, including differentiating cells of the gut and salivary glands. ARID3A represents a member of a unique family of transcriptional activators that shares sequence similarity to proteins of SWI/SNF complexes; it contains an A/T-rich DNA-binding (ARID) domain and a distinct domain involved in tetramerization. The gene encoding ARID3A is linked to a marker of Peutz-Jeghers syndrome, which is an autosomal-dominant disorder characterized by melanocytic macules of the lips, multiple gastrointestinal hamartomatous polyps and an increased risk for various neoplasms, including gastrointestinal cancer. E2FBP1 (E2F-1 binding protein 1) is identical to ARID3A in the carboxy terminal region. E2FBP1 appears to lack DNA binding and transactivation domains, and it functions to regulate the transcription of proteins involved in cell proliferation by binding to the transcription factor E2F-1.

Type: Primary
Antigen: DRIL1/ARID3A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/593
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge