Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DMWD Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13042R-A488
BOSSBS-13042R-A488EA 893 EUR InStock
BOSSBS-13042R-A488
Anti-DMWD Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
DMWD is a 674 amino acid protein containing five WD repeats. DMWD may play a role in the development of mental symptoms in severe cases of myotonic dystrophy, a chronic multisystemic disease characterized by wasting of the muscles, heart conduction defects, cataracts, endocrine changes and myotonia. The DMWD gene is located upstream of the DMPK gene and is prominently expressed in tissues affected in myotonic dystrophy patients. DMWD may also contribute to regulation in meiosis. DMWD is expressed in kidney and spleen, with strongest expression in brain, liver and testis. The gene encoding DMWD maps to human chromosome 19q13.32 and mouse chromosome 7 A3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DMWD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge