Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DYX1C1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13043R
BOSSBS-13043REA 725 EUR InStock
BOSSBS-13043R
Anti-DYX1C1 Rabbit Polyclonal Antibody
Antikörper
The gene encoding DYX1C1 maps in the 15q21 region, which is disrupted by a translocation t(2;15)(q11;q21) and segregates with dyslexia. Two sequence changes in DYX1C1, including one involving the translation initiation sequence and an Elk-1 transcription factor binding site (-3G --> A) and a codon (1249G --> T), introduce a premature stop codon and truncate the protein by 4 amino acids. DYX1C1 encodes a nuclear tetratricopeptide repeat domain protein that is dynamically regulated in brain. In human brain, DYX1C1 protein localizes to a fraction of cortical neurons and white matter glial cells. It is also expressed in lung, kidney and testis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DYX1C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge