Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EEF1D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13055R-FITC
BOSSBS-13055R-FITCEA 900 EUR InStock
BOSSBS-13055R-FITC
Anti-EEF1D Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
This gene encodes a subunit of the elongation factor-1complex, which is responsible for the enzymatic delivery ofaminoacyl tRNAs to the ribosome. This subunit, delta, functions asguanine nucleotide exchange factor. It is reported that followingHIV-1 infection, this subunit interacts with HIV-1 Tat. Thisinteraction results in repression of translation of host cellproteins and enhanced translation of viral proteins. Severalalternatively spliced transcript variants encoding multipleisoforms have been found for this gene. Related pseudogenes havebeen defined on chromosomes 1, 6, 7, 9, 11, 13, 17, 19.[provided byRefSeq, Aug 2010].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EEF1D
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge