Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EFEMP2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13059R-CY5
BOSSBS-13059R-CY5EA 900 EUR InStock
BOSSBS-13059R-CY5
Anti-EFEMP2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Fibulin proteins contribute to normal development of elastic fiber systems in various types of organs that require elasticity, such as vasculature, lung and skin. Fibulin-4, also known as EFEMP2 (EGF-containing fibulin-like extracellular matrix protein 2), MBP1 or UPH1 is a 443 amino acid secreted protein that contains six EGF-like calcium-binding domains and belongs to the fibulin family. Expressed ubiquitously with highest expression in heart, Fibulin-4 is essential for connective tissue development and elastic fiber formation, and may also play an important role in vascular patterning and collagen biosynthesis. Defects in the gene encoding Fibulin-4 are associated with autosomal recessive cutis laxa type I (CL type I), a connective tissue disorder that is inherited in both an autosomal dominant and an autosomal recessive manner and is characterized by inelastic tissue in all affected areas of the body.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EFEMP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge