Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EGFL6 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13062R-CY5
BOSSBS-13062R-CY5EA 900 EUR InStock
BOSSBS-13062R-CY5
Anti-EGFL6 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This gene encodes a member of the epidermal growth factor(EGF) repeat superfamily. Members of this superfamily arecharacterized by the presence of EGF-like repeats and are ofteninvolved in the regulation of cell cycle, proliferation, anddevelopmental processes. The gene product contains a signalpeptide, suggesting that it is secreted; an EGF repeat regionconsisting of 4 complete EGF-like repeats and 1 partial EGF-likerepeat, 3 of which have a calcium-binding consensus sequence; anarg-gly-asp integrin association motif; and a MAM domain, which isbelieved to have an adhesive function. This gene is expressed earlyduring development, and its expression has been detected in lungand meningioma tumors. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EGFL6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge