Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EIF1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13066R-HRP
BOSSBS-13066R-HRPEA 900 EUR InStock
BOSSBS-13066R-HRP
Anti-EIF1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
In mammalian cells, translation is controlled at the level of polypeptide chain initiation by initiation factors. Eukaryotic translation initiation factor 1 (eIF1) is crucial for the scanning process in vitro. During the scanning process, eIF1 is a component of a complex involved in recognition of the initiator codon. Translation is also initiated by the role of eIF1 in regulating the activity of ribosomal subunits 43S, 48S and 40S. eIF1 enables 43S ribosomal complexes to discern between cognate and near-cognate initiation codons, sensing the nucleotide content of initiation codons. It is also a promotor, along with eukar-yotic translation initiation factor 1A (eIF1A), for assembly of 48S ribosomal complexes at the initiation codon of a conventional capped mRNA. In addition, eIF1 and eIF1A, together with eukaryotic translation initiation factor 5 (eIF5), function in the formation of stable 40S ribosomal preinitiation complexes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: EIF1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge