Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ERF/Ets2 Repressor Factor Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13096R-A750
BOSSBS-13096R-A750EA 890 EUR InStock
BOSSBS-13096R-A750
Anti-ERF/Ets2 Repressor Factor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
ABT1 (activator of basal transcription 1) is a nuclear protein that associates with the TATA-binding protein (TBP) and enhances basal transcription activity of class II promoters. ABT1 associates with TBP in HeLa nuclear extracts in vitro. Another protein, designated ERF, is a member of the Ets family of transcription factors. The members of the Ets family are grouped because they share a highly conserved DNA binding domain. These factors are involved in growth factor pathways and regulate both proliferation and differentiation. ERF (Ets-2 repressor factor) is a ubiquitously expressed Ets-domain protein that exhibits strong transcriptional repressor activity, suppresses Ets-induced transformation and is regulated by MAPK phosphorylation. ERF transcription may be regulated by Ets-domain proteins. Additionally, modulation of ERF activity is involved in the transcriptional regulation of genes activated during entry into G1 phase.

Type: Primary
Antigen: ERF/Ets2 Repressor Factor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-160/548
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge