Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ERp19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13105R-A750
BOSSBS-13105R-A750EA 890 EUR InStock
BOSSBS-13105R-A750
Anti-ERp19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Endoplasmic reticulum proteins (ERps) are widely expressed proteins and localize to the ER. ERp19, ERp29, ERp46, ERp57 and ERp72 may act as proteases, protein disulfide isomerases, thiol-disulfide oxidases, phospholipases or a combination of these. ERp19, also designated thioredoxin domain-containing protein 12 (TXNDC12), and ERp46, also designated thioredoxin domain containing 5 (TXNDC5), belong to the thioredoxin superfamily and contain a thioredoxin fold with a consensus active-site sequence (CxxC). Both ERp19 and ERp46 are widely expressed ER luminal proteins that are most abundant in the liver and are enriched in purified liver ER vesicles. ERp19 shows significant protein thiol-disulfide oxidase activity in vitro, which is dependent on the presence of both active-site cysteines.

Type: Primary
Antigen: ERp19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/172
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge