Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EXOSC7 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13122R-CY7
BOSSBS-13122R-CY7EA 900 EUR InStock
BOSSBS-13122R-CY7
Anti-EXOSC7 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The exosome is a multisubunit complex of 3’ to 5’ exoribonucleases. It is involved in a variety of cellular processes and is responsible for degrading unstable mRNAs that contain AU-rich elements in their untranslated 3’ region. EXOSC7 (exosome component 7), also known as p8, EAP1, RRP42 (Ribosomal RNA-processing protein 42), Rrp42p or hRrp42p, is a component of the exosome multienzyme ribonuclease complex. It belongs to the RNase PH family and localizes to the nucleolus. EXOSC7 is one of the six RNase-PH domain subunits of the exosome. Together, these six subunits form a PNPase-like ring. EXOSC7 is required for the processing of the 7S pre-RNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EXOSC7
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge