Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EXTL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13123R-A488
BOSSBS-13123R-A488EA 893 EUR InStock
BOSSBS-13123R-A488
Anti-EXTL3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
EXTL3 is a member of the EXT (hereditary multiple exostosin) gene family of tumor suppressors encoding glycosyltransferases involved in heparan sulfate (HS) biosynthesis. Within this family, the C-terminus is conserved between all members from C. elegans to vertebrates. EXTL3 is a ubiquitously expressed, developmentally regulated, single-pass type II membrane protein that localizes to the endoplasmic reticulum membrane. EXTL3 adds N-acetylglucosamine (GlcNAc) to the polysaccharide-protein linkage region and to the growing HS chain suggesting that it plays a role in both the initiation and elongation of HS chains. In addition, EXTL3 may act as a Reg receptor, binding Reg via its N-terminus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EXTL3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge