Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EYA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13125R-HRP
BOSSBS-13125R-HRPEA 900 EUR InStock
BOSSBS-13125R-HRP
Anti-EYA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
A gene on chromosome 20q13.1 encodes Eya2 (eyes absent). EYA2 is one of four members of the eyes absent family. A 271 amino acid domain at the carboxy-terminal is highly conserved amongst the members of the eyes absent family, while the PST (proline-serive-threonin)-rich amino-terminal is highly divergent. EYA2 is expressed relatively late in development in the cytoplasm of extensor tendons and ligaments of the phalangeal elements of the limb, cranial placodes, branchial arches, central nervous system, and the developing eye. Pax3 induces the expression of Eya2 in a cascade that is necessary and sufficient for myogenesis. EYA2, like EYA1, acts as a transcriptional activator in connective tissue patterning through its PST domain, which functions as a transactivation domain. EYA2 is translocated to the nucleus by Six proteins, which interact through their domain and homeodomain with EYA2. EYA2 carboxy-terminal interacts with the G Alpha z and G Alphai 2 proteins. This interaction prevents Six proteins from translocating EYA2 to the nucleus.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: EYA2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge