Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FANCC Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13140R-FITC
BOSSBS-13140R-FITCEA 900 EUR InStock
BOSSBS-13140R-FITC
Anti-FANCC Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Fanconi anemia (FA) is an autosomal recessive disorder characterized by bone marrow failure, birth defects and chromsomal instability (1,2). The FA Group C complementation group gene encodes the protein FANCC, which is located in both cytoplasmic and nuclear compartments. FANCC is expressed in a cell cycle-dependent manner, with the lowest levels at the G1/S boundary and the highest levels in the M-phase. The FANCC protein interacts with other FA complementation group proteins as well as non-FA proteins (3). A human a spectrin II (designated aSpIIs) acts as a scaffold to enhance interactions between FANCC and FANCA to form a nuclear complex (4,5). Another binding partner of FANCC is the BTB/POZ domain containing protein FAZF, which is a transcriptional repressor (6). In hematopoietic cells expressing mutant FANCC, PKR is constitutively phosphorylated and has increased binding affinity for double-stranded RNA (7,8), which suggests that FANCC indirectly suppresses the activity of PKR. These cells are also apoptotic and are hypersensitive to IFNg and TNFa (8). In addition, FANCC protein is involved in the activation of STAT1 through receptors for at least three hematopoietic growth and survival factors (8).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FANCC
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge