Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FANCM Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13143R-CY3
BOSSBS-13143R-CY3EA 900 EUR InStock
BOSSBS-13143R-CY3
Anti-FANCM Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Fanconi anemia (FA) is an autosomal recessive disorder characterized by bone marrow failure, birth defects and chromosomal instability. At the cellular level, FA is characterized by spontaneous chromosomal breakage and a unique hypersensitivity to DNA cross-linking agents. The thirteen FA proteins that have been characterized are important for regulating chromosomal stability and genome surveillance. Eight of these proteins, namely FANCA, FANCB, FANCC, FANCE, FANCF, FANCG, FANCL and FANCM, comprise the FA core complex, which catalyzes a key reaction in DNA repair: the monoubiquitination of FANCD2. FANCM (Fanconi anemia, complementation group M) is a member of the DEAD-box helicase family of proteins and contains a DEAH helicase domain and a nuclease domain. Localizing to chromatin fractions, FANCM is phosphorylated in a cell cycle-dependent manner and is believed to function as an anchor, recruiting the FA core complex to chromatin. Mutations in the gene encoding FANCM can lead to Fanconi anemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FANCM
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge