Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FARP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13145R-A488
BOSSBS-13145R-A488EA 893 EUR InStock
BOSSBS-13145R-A488
Anti-FARP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Aminoacyl-tRNA synthetases consist of a family of enzymes that catalyze the specific aminoacylation of tRNA by their cognate amino acid in the initial step of ribosome-dependent protein biosynthesis. FARSLA, also known as FRSA, CML33, FARSL or PheHA (phenylalanyl-tRNA synthetase, alpha subunit), is a member of the class-II aminoacyl-tRNA synthetase family and is highly expressed in proliferating cells of bone marrow. FARSLA is a cytoplasmic phenylalanine-tRNA synthetase that functions as a heterodimer consisting of a catalytic alpha-subunit and a regulatory beta-subunit. The alpha-subunit is responsible for forming the amino acid binding pocket, mediating the ATP/aminoacyl adenylate binding, and interacts with the acceptor stem of the tRNA. FARSLA functions in a cell cycle-dependent and differentiation-dependent manner.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FARSLA/CML33
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge