Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FAU Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13148R-FITC
BOSSBS-13148R-FITCEA 900 EUR InStock
BOSSBS-13148R-FITC
Anti-FAU Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates. The first step requires the ATP-dependent activation of the Ub C-terminus and the assembly of multi-Ub chains by the Ub-activating enzyme known as the E1 component. The Ub chain is then conjugated to the Ub-conjugating enzyme (E2) to generate an intermediate Ub-E2 complex. The Ub-ligase (E3) then catalyzes the transfer of Ub from E2 to the appropriate protein substrate. A wide range of enzymes facilitate in the proteolytic Ub pathway, including monoclonal nonspecific suppressor factor-beta (MNSF-beta), a subunit of MNSF, which is a lymphokine product of a murine T cell hybrid-oma that restricts the production of LPS-induced immunoglobulin secreting cells in an antigen-nonspecific manner. MNSF-beta is a ubiquitin-like fusion protein consisting of the ribosomal protein S30 and a protein that shares 36% sequence identity with ubiquitin. This ubiquitin-like segment (Ubi-L) can be cleaved from MNSF-beta in the cytosol.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAU/MNSF-beta
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge