Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FBLN7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13161R-A350
BOSSBS-13161R-A350EA 893 EUR InStock
BOSSBS-13161R-A350
Anti-FBLN7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Fibulin-7 is a 439 amino acid extracellular matrix protein that belongs to the Fibulin family. Containing two EGF-like domains and one sushi (CCP/SCR) domain, Fibulin-7 exists as four alternatively spliced isoforms. Fibulin-7 is considered an adhesion protein that interacts with extracellular matrix molecules in developing teeth, and may be involved in differentiation and maintenance of odontoblasts as well as in dentin formation. Fibulin-7 is post-translationally glycosylated with N-linked oligosaccharides and interacts with heparin, fibronectin, fibulin-1 and DSP (dentin sialophosphoprotein). Fibulin-7 is encoded by a gene located on human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fibulin 7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge