Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FCN2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13162R-CY5
BOSSBS-13162R-CY5EA 900 EUR InStock
BOSSBS-13162R-CY5
Anti-FCN2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Ficolin B is the designation in mouse and rat of a protein also known as L-ficolin, ficolin-2, collagen/fibrinogen domain-containing protein 2, serum lectin p35, EBP-37 or hucolin. Ficolin B is a 313 amino acid member of the ficolin lectin family. It is a secreted innate immunity pattern recognition molecule expressed in liver and plasma that forms a disulfide-linked homopolymer. This extensive N-terminal disulfide bridge formation can lead to a functional dodecamer polypeptide. Ficolin B binds to DNA ligands expressed by late apoptotic and necrotic cells to increase attachment and engulfment. Variation in ficolin B concentrations amongst individuals is associated with polymorphisms in the promoter and structural portion of the FCN2 gene. In patients with Beh鏴t's disease (BD), there exists a significant difference in allele frequency for FCN2 gene single nucleotide polymorphisms (SNPs) within the -557 and -64 promoter sites within HLA-B51 positive and HLA-B51 negative subgroups.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Ficolin 2/Ficolin B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge