Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FIP1L1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13173R-FITC
BOSSBS-13173R-FITCEA 900 EUR InStock
BOSSBS-13173R-FITC
Anti-FIP1L1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
The Component of the Cleavage and Polyadenylation Specificity Factor (CPSF) complex plays an important role in the 3'-end formation of pre-mRNA. This complex recognizes the AAUAAA signal sequence and interacts with poly(A) polymerase to process and add to the poly(A) tail. FIP1L1 (FIP1-like 1), also known as Pre-mRNA 3'-end-processing factor FIP1, FIP1 (Factor interacting with PAP) and RHE (Rearranged in hypereosinophilia), is a 594 amino acid nuclear protein that is a component of the CPSF complex. Within the complex, FIP1L1 contributes to the poly(A) recognition and stimulates poly(A) addition. Fusion of the genes encoding FIP1L1 and PDGFRA due to an interstitial deletion on chromosome 4q12 is the cause of hypereosinophilia syndrome, a rare blood disorder characterized by continuous overproduction of eosinophils in the bone marrow that leads to tissue infiltration and organ damage. There are three isoforms of FIP1L1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FIP1L1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge