Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FKBP11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13176R-A488
BOSSBS-13176R-A488EA 893 EUR InStock
BOSSBS-13176R-A488
Anti-FKBP11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The immunophilins are a highly conserved family of cis-trans peptidyl-prolyl isomerases that bind to and mediate the effects of immunosuppressive drugs, such as cyclosporin, FK506 and rapamycin. Immunophilins have also been implicated in protein folding and trafficking within the endoplasmic reticulum (ER). FKBP11 (FK506-binding protein 11), also known as FKBP19 or peptidyl-prolyl cis-trans isomerase FKBP11, is a 201 amino acid single-pass membrane protein belonging to the FKBP-type PPIase family, a group of proteins known to catalyze the folding of proline-containing polypeptides. Containing one PPIase FKBP-type domain, FKBP11 is expressed in secretory tissues such as pancreas, pituitary, stomach, lymph node and salivary gland, and is encoded by a gene that maps to human chromosome 12q13.12. FK506 and rapamycin are known to inhibit FKBP11’s peptidyl-prolyl isomerase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FKBP11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge