Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FMO3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13186R-A488
BOSSBS-13186R-A488EA 893 EUR InStock
BOSSBS-13186R-A488
Anti-FMO3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The Flavin containing monooxygenase family consists of five gene products, FMO1-5, that are major enzymatic oxidants involved in the metabolism of various therapeutics. Located in the liver, FMO3 is a hepatic microsomal enzyme that oxygenates soft nucleophiles such as secondary and tertiary amines. Through its N-oxygenase capabilities, FMO3 acts on a variety of xenobiotics to catalyze oxidative digestion. Defects in the FMO3 gene are the primary cause of trimethylaminuria (TMAuria), an inborn error of metabolism associated with a fishy body odor emitting from sweat, urine and breath. Genetic mutations in FMO3 lead to the N-oxidation of amino-trimethylamine derived from food products, thus producing the malodor associated with TMAuria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FMO3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge