Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FN3K Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13189R-A680
BOSSBS-13189R-A680EA 890 EUR InStock
BOSSBS-13189R-A680
Anti-FN3K Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Amines, including those present on proteins, spontaneously react with glucose to make fructosamines in a reaction termed glycation. Fructosamine 3-kinase (FN3K), a 309-amino acid enzyme initially identified in erythrocytes, catalyzes the ATP-dependent phosphorylation of the third carbon on both D- and L-fructosamines, leading to their destabilization and eventually, their removal from the protein. FN3K is a monomer that is ubiquitously expressed in mammalian tissue and phosphorylates both low molecular mass and protein-bound fructosamines which are formed as a result of glycation of glucose with primary amines. FN3K protects proteins from the harmful effects of nonenzymatic glycation, and may also be involved in peptide repair and cell metabolism. FN3KRP (fructosamine-3-kinase-related protein) is a 309 amino acid protein that is expressed in erythrocytes, bone marrow, spleen, brain and kidney and belongs to the fructosamine kinase family. FN3KRP functions to phosphorylate psicoamines and ribulosamines on the third carbon of their sugar moiety, thereby leading to the deglycation of the target amines.

Type: Primary
Antigen: FN3K
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-309/309
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge