Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FNDC3A Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13191R-A750
BOSSBS-13191R-A750EA 890 EUR InStock
BOSSBS-13191R-A750
Anti-FNDC3A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
FNDC3A is a 1,134 amino acid protein that belongs to the FNDC3 family of proteins. FNDC3A contains an N-terminal proline-rich region, nine fibronectin type-III domains (none of which contain an RGD sequence) and a hydrophobic C-terminal transmembranous helix. Expressed in a wide variety of tissues, FNDC3A localizes to Golgi vesicles and to the developing acrosome of spermatids. FNDC3A is believed to function in glycosaminoglycan and collagen synthesis. In mice, a mutation in the gene encoding FNDC3A causes male sterility due to defective adhesion between Sertoli cells and spermatids in the seminiferous epithelium. This suggests that FNDC3A plays an important role in spermatogenesis, possibly mediating or maintaining the adhesion between Sertoli cells and spermatids.

Type: Primary
Antigen: FNDC3A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/1198
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge