Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FNDC4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13192R-CY7
BOSSBS-13192R-CY7EA 900 EUR InStock
BOSSBS-13192R-CY7
Anti-FNDC4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Fibronectins are multi-domain glycoproteins that bind to a variety of substances including collagen, actin, heparin, DNA, fibrin and fibronectin receptors. They are involved in a diverse array of important functions such as blood coagulation, wound healing, cell adhesion, cell differentiation and migration. FNDC4 (Fibronectin type III domain-containing protein 4), also known as FRCP1 (Fibronectin type III repeat-containing protein 1), is a 234 amino acid membrane protein that contains one fibronectin type-III domain, which serves as a binding site for DNA, heparin or the cell surface. The gene encoding FNDC4 is localized to human chromosome two, which houses over 1,400 genes and comprises nearly 8% of the human genome.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FNDC4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge