Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXH1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13202R-CY3
BOSSBS-13202R-CY3EA 900 EUR InStock
BOSSBS-13202R-CY3
Anti-FOXH1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Xenopus winged-helix factor, xFAST-1 (forkhead activin signal transducer-1) is a transcription factor that forms a complex with the receptor-regulated Smad protein, Smad2, and directly binds to activin response elements on DNA (1,2). The human homolog FAST-1 and the corresponding mouse homolog, designated FAST-2, share significant sequence homology with xFAST-1, including a conserved N-terminal forkhead domain that consists of 110 amino acid residues and is essential for binding DNA and regulating transcription in embryogenesis, in tumorigenesis and in the maintenance of differentiated cell states (3,4). FAST-1 and FAST-2 also contain a distinct C-terminal Smad interaction domain that is required for the association with various Smad proteins, including Smad2, Smad3 and Smad4 (3,5). Expression of FAST-1 and FAST-2 is predominantly observed during early development, with lower levels detected in adult tissues (6,7). FAST-1 and FAST-2 mediated DNA binding is attenuated by both TFGß and activin, indicating that these FAST proteins mediate TFGß induced signal transduction (3).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXH1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge