Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXI1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13203R-A647
BOSSBS-13203R-A647EA 893 EUR InStock
BOSSBS-13203R-A647
Anti-FOXI1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
FOXI1 is a member of the FOX family of transcription factors. The FOX family is a large group of proteins (consisting of at least 43 members) that share a common DNA binding domain termed winged-helix or forkhead domain. FOX transcription factors play important roles in development, differentiation, aging and hormone responsiveness. Localizing to the nucleus, FOXI1 functions as a transcription factor. Mice with mutated forms of FOXI1 show defects in ear development, implying that FOXI1 plays a significant role in the developmental pathway of ears and, in particular, the cochlea and vestibulum. FOXI1 is an upstream transcription regulator of Pendrin (a protein associated with deafness), suggesting a role for FOXI1 in the pathogenesis of Pendred syndrome (PS), a condition of nonsyndromic hearing loss and enlarged vestibular aqueduct (EVA).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXI1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge