Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXN1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13205R-CY3
BOSSBS-13205R-CY3EA 900 EUR InStock
BOSSBS-13205R-CY3
Anti-FOXN1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The forkhead domain-containing gene family (Fox) comprises over 20 members in mammals and is defined by a conserved 110 amino-acid motif containing a winged helix structure DNA-binding domain. The members of this gene family have been implicated as key regulators of embryogenesis, cell cycling, cell lineage restriction and cancer. As such, FOXN2 contains a domain with homology to the forkhead DNA binding domain. FOXN2, or Human T-cell leukemia virus enhancer factor, is a 341 amino acid protein mapping to human gene FOXN2, which has been localized to human chromosome 2p16-p22. This protein, encoded by a 1239-bp cDNA isolated from the Jurkat cDNA library, is capable of binding to a region of the human T-cell leukemia virus long terminal repeat (HTLV-I LTR) located between amino acids 155 and 117. This purine-rich region is important in the regulation of gene expression by the Ets family of transcription factors. FOXN2 is a unique cellular gene that may function in the transcriptional regulation of HTLV-I LTR.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXN2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge