Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXRED1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13209R-FITC
BOSSBS-13209R-FITCEA 900 EUR InStock
BOSSBS-13209R-FITC
Anti-FOXRED1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
FOXRED1 is a 486 amino acid single-pass membrane protein. Utilizing FAD as a cofactor, FOXRED1 may act as a chaperone protein essential for the function of mitochondrial complex I. Mutations to FOXRED1 may result in mitochondrial complex I deficiency (MT-C1D), which results in a wide range of clinical maladies from lethal neonatal disease to adult onset neurodegenerative disorders. Common phenotypes of MT-C1D include cardiomyopathy, liver disease, Leigh syndrome, Leber hereditary optic neuropathy, and some forms of Parkinson disease. FOXRED1 exists as three alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 11q24.2. With approximately 135 million base pairs and 1,400 genes, chromosome 11 makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXRED1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge